Order Entry
Germany
ContactUsLinkComponent
Ampicillin trihydrate, Supelco®
Ampicillin trihydrate, Supelco®
Catalog # 31591-250MG
Supplier:  Merck
CAS Number:  
undefined
Ampicillin trihydrate, Supelco®
Catalog # 31591-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :403.45 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:200-709-7
 :GHS07
Storage Temperature:Refrigerator
CAS Number:7177-48-2
Formula:C₁₆H₁₉N₃O₄S·3H₂O
MDL Number:MFCD00072036