Order Entry
Germany
ContactUsLinkComponent
 
Ampicillin trihydrate
Catalog # PROOMM0253.15
CAS Number:  
Ampicillin trihydrate
Catalog # PROOMM0253.15
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :403.45 g/mol
EINECS:200-709-7
 :GHS07
Storage Temperature:Refrigerator
CAS Number:7177-48-2
Formula:C₁₆H₁₉N₃O₄S·3H₂O
MDL Number:MFCD00072036