Order Entry
Germany
ContactUsLinkComponent
Ampicillin trihydrate
Irritant
Ampicillin trihydrate
Catalog # MOLE14575910-25G
Supplier:  Molekula
CAS Number:  
Irritant
Ampicillin trihydrate
Catalog # MOLE14575910-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :403.45 g/mol
EINECS:200-709-7
 :GHS07
Storage Temperature:Refrigerator
CAS Number:7177-48-2
Formula:C₁₆H₁₉N₃O₄S·3H₂O
MDL Number:MFCD00072036