Buffer, PBS solution 25X concentrate (phosphate buffered saline)
PBS buffer solution 25X concentrate (phosphate buffered saline), Glass bottle, 3.000 mL
Catalog # TREV4868-500-06
Marketsource
Supplier: TREVIGEN
Buffer, PBS solution 25X concentrate (phosphate buffered saline)
Catalog # TREV4868-500-06
Supplier: TREVIGEN
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:3.000 mL
- Description:PBS buffer solution 25X concentrate (phosphate buffered saline)



