Order Entry
Germany
ContactUsLinkComponent
3-Amino-1H-1,2,4-triazole-5-thiol
3-Amino-1H-1,2,4-triazole-5-thiol
Catalog # MOLE20831626-25G
Supplier:  Molekula
CAS Number:  
3-Amino-1H-1,2,4-triazole-5-thiol
Catalog # MOLE20831626-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :116.15 g/mol
EINECS:240-735-6
CAS Number:16691-43-3
Formula:C₂H₄N₄S
MDL Number:MFCD00005231