Kanamycin sulphate hydrate, min. 750 I.U./mg, BioChemica
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:5965-95-7
:25389-94-0
Formula:C₁₈H₃₆N₄O₁₁·H₂SO₄·H₂O
:600.60 g/mol
EINECS:246-933-9
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:min. 750 I.U./mg
- α20°C/D; 1 %, H2O:+112° - +123°
- pH (1 %; H2O):6.5 - 8.5
- Loss on drying (60°C):max. 1.5 % (1 Torr)
- Kanamycin B:max. 4 %