Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Ampicillin

Marketsource
Supplier:   Serva
Ampicillin
Supplier:  Serva
Marketsource
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:69-53-4
  • Formula:C₁₆H₁₉N₃O₄S
  • MDL Number:MFCD00005175
  • Molecular Weight:349.4 g/mol
  • EINECS:200-709-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 67,60 €
    67,60 €
    1 * 10 g
       67,60 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 408,00 €
    408,00 €
    1 * 100 g
       408,00 €

    Specifications:
    Marketsource
    Marketsource