Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Ampicillin

Glass bottle for solids, 500 mg
Catalog # PROOMM0253.16
Ampicillin
Catalog # PROOMM0253.16
CAS Number:  69-53-4
Ampicillin

Glass bottle for solids, 500 mg

CAS Number:  69-53-4
Price
1 240,00 €
Log in for availability
1 * 500 mg
Product Details & Documents
  • CAS Number:69-53-4
  • Formula:C₁₆H₁₉N₃O₄S
  • MDL Number:MFCD00005175
  • Molecular Weight:349.4 g/mol
  • EINECS:200-709-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications