Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Amidosulphonic acid
Amidosulphonic acid 10% in aqueous solution, extra pure
Amidosulphonic acid 10% in aqueous solution, extra pure
Amidosulphonic acid 10% in aqueous solution, extra pure
Supplier:
Bernd Kraft
Certificates
Expand image
Amidosulphonic acid 10% in aqueous solution, extra pure
Supplier:
Bernd Kraft
About this item
CAS Number:
5329-14-6
:
Formula:
H₃NO₃S
MDL Number:
MFCD00011603
:
97.09 g/mol
EINECS:
226-218-8