Hand protectors, Hot Hand®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
This protector is made of durable silicone rubber which remains flexible from –57 to +260 °C.
- Safe, non-slip protection against very hot or cold surfaces
- End pockets accept thumb and fingers to grasp objects
- Multiple studs with concave tips cover surface of the Mitt, providing secure grip on virtually all hot/cold surfaces
- One size fits all hands, left or right
Dimensions: 100×190 mm