ContactUsLinkComponent
Sampling knives, disposable, Sterileware®, SP Bel-Art
Catalog # 231-0016
undefined
Sampling knives, disposable, Sterileware®, SP Bel-Art
Catalog # 231-0016
Specifications
  • Type:
    Serrated and smooth edge blade
  • Total length:
    209 mm
  • Pk:
    200
Specifications
About this item
PS, white.
  • Ideal for food sampling, e.g. meats, vegetables, seafood and other fibrous and semi solid materials
  • All knives are lot traceable
  • Jagged blade cuts efficiently, opposite edge can spread material
  • Individually packaged and sterilised
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904