VWR®, DNA ladder, 100 bp
K180-250UL
:
- Pk:250 µl
- Description:100 bp ladder, sufficient for 50 lanes
Ladder with 13 fragments ranging from 100 to 3000 bp for easy band identification.
Also available pre-mixed with loading dye.
Ladder with 13 fragments ranging from 100 to 3000 bp for easy band identification.
Also available pre-mixed with loading dye.