ContactUsLinkComponent
Tubing cutter, BOLA
Tubing cutter, BOLA
Catalog # BOHLS1852-28
Supplier:  Bohlender
undefined
Tubing cutter, BOLA
Catalog # BOHLS1852-28
Supplier:  Bohlender
Specifications
  • Description:
    Tubing cutter, for plastic and rubber tubing upto Ø 28 mm, not suitable for steel reinforced tubing
  • Pk:
    1
Specifications
About this item
Ideal for cutting plastic and rubber tubing with and without textile reinforcement.
  • Blade is exchangeable
  • Not suitable for steel reinforced tubing