EDTA, 0.5 M solution, pH 8.0, for biotechnology
Supplier: VWR Chemicals
Certificates
Frequently Bought Together
Jerricans, narrow neck, HDPE, transparent or blue
Supplier: Kautex
U96 DeepWell™ plates 1,3 and 2,0 ml, Nunc®
Supplier: Thermo Fisher Scientific
Water nuclease-free
Supplier: OMEGA BIO-TEK
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: Sterilin
EDTA, 0.5 M solution, pH 8.0, for biotechnology
Supplier: VWR Chemicals
VWR®, TE (Tris-EDTA) buffer, pH 8,0 for biotechnology
Supplier: VWR Chemicals
Hexylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Sodium chloride 5 mol/L (5 M) in aqueous solution for molecular biology
Supplier: PanReac AppliChem
Proteinase K solution
Supplier: PanReac AppliChem
Water, Cell Culture Grade pyrogen-free
Supplier: VWR Chemicals
VWR® Square Polycarbonate Media Bottles
Supplier: VWR Collection
VWR® Buffer, HEPES Solution pH 7,3 (1 mol/L)
Supplier: VWR Chemicals
Buffer, HEPES solution pH 7,3 (1 mol/L), Molecular Biology Grade, Ultrapure
Supplier: Thermo Fisher Scientific
Reagecon 251.3 mS/cm Conductivity Standard at 25C 1 * 500 mL
Supplier: REAGECON
b.safe Funnels 250
Supplier: Bohlender
Tris-HCl, 1M solution, pH 8,0, Molecular Biology Grade, Ultrapure
Supplier: Thermo Fisher Scientific
MRS BROTH, FOR MICROBIOLOGY 1 * 500 g
Supplier: Merck
Sodium dodecyl sulphate (SDS) 200 g/l in aqueous solution
Supplier: Serva