Order Entry
Germany
ContactUsLinkComponent
 
Cobalt standard dissolved in oil, 5,000 µg/g Co in Base Oil 75 for wear metal analysis
Catalog # JOBIORG-CO8-4Z
CAS Number:  
Cobalt standard dissolved in oil, 5,000 µg/g Co in Base Oil 75 for wear metal analysis
Catalog # JOBIORG-CO8-4Z
CAS Number:  

Specifications

  • Concentration:
    5000 µg/g
  • Matrix:
    Base Oil 75
  • Pk:
    1
  • Element:
    Cobalt
  • Symbol:
    Co

Specifications

About this item