Cobalt standard dissolved in oil, 1,000 µg/g Co in Base Oil 75 for wear metal analysis
Catalog # JOBIORG-CO8-2Z
Supplier: JOBIN YVON EMISSION
CAS Number:
Specifications
- Concentration:1000 µg/g
- Matrix:Base Oil 75
- Pk:1
- Element:Cobalt
- Symbol:Co