Order Entry
Germany
ContactUsLinkComponent
 
Aluminium standard dissolved in oil, 5000 µg/g Al in Base Oil 75 for wear metal analysis
  JOBIORG-AL8-4Z
 :  
Aluminium standard dissolved in oil, 5000 µg/g Al in Base Oil 75 for wear metal analysis
  JOBIORG-AL8-4Z
 :  

 

  • Concentration:
    5000 µg/g
  • Matrix:
    Base Oil 75
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Element:
    Aluminium
  • Symbol:
    Al