Order Entry
Germany
ContactUsLinkComponent
Vinclozolin
Vinclozolin
Catalog # EHERC17920000
Supplier:  EHRENSTORFER
CAS Number:  
Vinclozolin
Catalog # EHERC17920000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
EINECS:256-599-6
CAS Number:50471-44-8
Formula:C₁₂H₉Cl₂NO₃