Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Orotic acid monohydrate
Orotic acid monohydrate ≥98%
Orotic acid monohydrate ≥98%
:
Thermo Fisher Scientific
Orotic acid monohydrate ≥98%
:
Thermo Fisher Scientific
:
50887-69-9
:
:
C₅H₄N₂O₄·1H₂O
:
MFCD00149398
:
156.10 g/mol (anhy)
:
14,6876
:
>300 °C
:
200-619-8
:
GHS07
:
Ambient
msds_ALFAA18594
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years