Order Entry
Germany
ContactUsLinkComponent
Phenylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®
Phenylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL257125-200ML
Supplier:  Merck
CAS Number:  
Phenylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL257125-200ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    200 mL
  • Seq:
    2530

Specifications

About this item

 :
 :136.86 g/mol
ADR:4.3,II
 :M-Clarity™ quality level = MQ100
EINECS:202-868-8
UN:3399
CAS Number:100-59-4
Formula:C₆H₅ClMg
MDL Number:MFCD00000464