Order Entry
Germany
ContactUsLinkComponent
Pesticide reference standard, Permethrin (mixture of cis- and trans-isomeres) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Permethrin (mixture of cis- and trans-isomeres) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15990000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Permethrin (mixture of cis- and trans-isomeres) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15990000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    52645-53-1
  • Description:
    Permethrin (mixture of cis- and trans-isomeres) 10 µg/ml in cyclohexane
  • Formula:
    C₂₁H₂₀C₁₂O₃
  • Description:
    Permethri

Specifications

About this item