Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-Indoline-2-carboxylic acid
(±)-Indoline-2-carboxylic acid
(±)-Indoline-2-carboxylic acid
(±)-Indoline-2-carboxylic acid
Supplier:
Thermo Scientific
Certificates
Expand image
(±)-Indoline-2-carboxylic acid
Supplier:
Thermo Scientific
About this item
CAS Number:
78348-24-0
:
Formula:
C₉H₉NO₂
MDL Number:
MFCD00010635
:
163.18 g/mol
EINECS:
278-899-6