α-Cyclodextrine hydrate 98+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: α-Cyclodextrin hydrate, α-Cyclodextrin xhydrate
- CAS Number:51211-51-9
- Formula:C₃₆H₆₀O₃₀·xH₂O
- MDL Number:MFCD00150804
- Molecular Weight:972.86 g/mol (anhy)
- Melting Pt:278 °C (dec.)
- Storage Temperature:Ambient
- EINECS:233-007-4
3 Options Available Below
Product Details & Documents
Synonyms: α-Cyclodextrin hydrate, α-Cyclodextrin xhydrate
- CAS Number:51211-51-9
- Formula:C₃₆H₆₀O₃₀·xH₂O
- MDL Number:MFCD00150804
- Molecular Weight:972.86 g/mol (anhy)
- Melting Pt:278 °C (dec.)
- Storage Temperature:Ambient
- EINECS:233-007-4
Documents
ACRO22729
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤11 %
- Sulfated ash:≤0.1 %
- HPLC:≥98.0 % (ELSD) on dry substance
- Specific optical rotation:+145° to +151° (20°C, 589 nm) (c=2, water) (on dry substance)
- Heavy metals (as Pb):≤15 ppm
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



