Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(Methylthio)nicotinoyl chloride

2-(Methylthio)nicotinoyl chloride

Marketsource
Supplier:   Thermo Scientific
2-(Methylthio)nicotinoyl chloride
Supplier:  Thermo Scientific
Marketsource
Certificates
2-(Methylthio)nicotinoyl chloride

2 Options Available Below

Product Details & Documents
  • CAS Number:97936-43-1
  • Formula:C₇H₆ClNOS
  • MDL Number:MFCD00051678
  • Molecular Weight:187.65 g/mol
  • UN:3261
  • ADR:8,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.