Order Entry
Germany
ContactUsLinkComponent
Butralin (4-tert-Butyl-N-sec-butyl-2,6-dinitroaniline)
Butralin (4-tert-Butyl-N-sec-butyl-2,6-dinitroaniline)
Catalog # EHERC10910000
Supplier:  EHRENSTORFER
CAS Number:  
Butralin (4-tert-Butyl-N-sec-butyl-2,6-dinitroaniline)
Catalog # EHERC10910000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
EINECS:251-607-4
CAS Number:33629-47-9
Formula:C₁₄H₂₁N₃O₄