Order Entry
Germany
Orders LinkContactUsLinkComponent
Sulphacetamide

Sulphacetamide

Glass bottle for solids, 100 g
Catalog # MOLE34100925-100G
Marketsource
Supplier:   Molekula
Sulphacetamide
Catalog # MOLE34100925-100G
Supplier:  Molekula
Marketsource
CAS Number:  144-80-9
Sulphacetamide

Glass bottle for solids, 100 g

CAS Number:  144-80-9
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  N-((4-Aminophenyl)sulphonyl)acetamide
  • CAS Number:144-80-9
  • Formula:C₈H₁₀N₂O₃S
  • MDL Number:MFCD00066501
  • Molecular Weight:214.24 g/mol
  • EINECS:205-640-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.