Pesticide reference standard, Binapacryl (2-sec-Butyl-4,6-dinitrophenyl-3-methylcrotonate) 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL10590000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:485-31-4
- Description:Binapacryl (2-sec-Butyl-4,6-dinitrophenyl-3-methylcrotonate) 10 µg/ml in cyclohexane
- Formula:C₁₅H₁₈N₂O₆
- Description:Binapacry