Order Entry
Germany
ContactUsLinkComponent
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)
Catalog # 04-12-5007-25
CAS Number:  
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)
Catalog # 04-12-5007-25
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :125.56 g/mol
EINECS:227-139-1
CAS Number:5680-79-5
Formula:C₃H₈ClNO₂
MDL Number:MFCD00012870