Order Entry
Germany
ContactUsLinkComponent
Methomyl (1-(methylthio)ethylideneamino-N-methylcarbamate)
Methomyl (1-(methylthio)ethylideneamino-N-methylcarbamate)
Catalog # MOLEM47421769
Supplier:  Molekula
CAS Number:  
Methomyl (1-(methylthio)ethylideneamino-N-methylcarbamate)
Catalog # MOLEM47421769
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
EINECS:240-815-0
UN:2757
ADR:6.1,II
CAS Number:16752-77-5
Formula:C₅H₁₀N₂O₂S