Order Entry
Germany
ContactUsLinkComponent
 
Dipropyleneglycol monomethyl ether (mixture of isomers) for synthesis, Sigma-Aldrich®
 :  Merck
    
 
Dipropyleneglycol monomethyl ether (mixture of isomers) for synthesis, Sigma-Aldrich®
 :  Merck
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    148.2 g/mol
  • Boiling Point:
    184 °C (1013 hPa)
  • Vapor Pressure:
    0.75 hPa (25 °C)
  • Flash Point:
    79 °C
  • Refractive Index:
    1.419 (25 °C, 589 nm)
  • Explosion Limit:
    1.1 - 14 %(V)
  • Density:
    0.95 g/cm3 (20 °C)
  • pH:
    6 - 7 (200 g/l, H2O, 20 °C)
  • Ignition Point:
    270 °C

 

 

Frequently Bought Together