Pesticide reference standard, Omethoate (O,O-Dimethyl S-methylcarbamoylmethyl phosphorothioate) 100 µg/ml in acetone, Dr Ehrenstorfer
EHERXA15730000AC
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:1113-02-6
- Description:Omethoate (O,O-Dimethyl S-methylcarbamoylmethyl phosphorothioate) 100 µg/ml in acetone
- Formula:C₅H₁₂NO₄PS
- Description:Omethoate (O,O-Dimethyl S-methylcarbamoylmethyl phosphorothioate)