Phosphorus standard solution, 1000 mg/L P in 0,5 M hydrochloric acid (from H₃PO₄) standard for AAS
Catalog # KRAF03367.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HCl
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Phosphorus
- Symbol:P
Specifications
About this item
Certifications: Traceable to NIST.