Neodymium standard solution, 1000 mg/L Nd in 0,5 M hydrochloric acid (from Nd₂O₃) standard for ICP
Catalog # KRAF21424.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Neodymium
- Symbol:Nd
Specifications
About this item
Certifications: Traceable to NIST