Order Entry
Germany
ContactUsLinkComponent
Calcium standard solution, 10000 mg/L Ca in 5% hydrochloric acid (from Ca) standard for AAS and ICP
Calcium standard solution, 10000 mg/L Ca in 5% hydrochloric acid (from Ca) standard for AAS and ICP
Catalog # JOBIPLCA1-3X
CAS Number:  
Calcium standard solution, 10000 mg/L Ca in 5% hydrochloric acid (from Ca) standard for AAS and ICP
Catalog # JOBIPLCA1-3X
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HCl
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item

Certifications: Traceable to SRM from NIST