Order Entry
Germany
ContactUsLinkComponent
 
Mercury standard solution, 10,000 mg/l Hg in water (from HgCl2) standard for AAS
Catalog # KRAF17213.0000
Supplier:  Bernd Kraft
CAS Number:  
Mercury standard solution, 10,000 mg/l Hg in water (from HgCl2) standard for AAS
Catalog # KRAF17213.0000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item