Silver standard solution, 10000 mg/L Ag in water (from K(Ag(CN)₂)) standard for AAS
Catalog # KRAF14114.0700
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:H₂O
- Pack type:Glass bottle
- Pk:100 mL
- Element:Silver
- Symbol:Ag