Order Entry
Germany
ContactUsLinkComponent
Magnesium standard solution, 10000 mg/L Mg in 5% hydrochloric acid (from Mg) standard for AAS and ICP
Magnesium standard solution, 10000 mg/L Mg in 5% hydrochloric acid (from Mg) standard for AAS and ICP
Catalog # JOBIPLMG1-3X
CAS Number:  
Magnesium standard solution, 10000 mg/L Mg in 5% hydrochloric acid (from Mg) standard for AAS and ICP
Catalog # JOBIPLMG1-3X
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    5% HCl
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Certifications: Traceable to SRM from NIST