Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Nitro-L-tyrosine
3-Nitro-L-tyrosine
3-Nitro-L-tyrosine
3-Nitro-L-tyrosine
Supplier:
Cayman Chemical
Certificates
Expand image
3-Nitro-L-tyrosine
Supplier:
Cayman Chemical
About this item
CAS Number:
621-44-3
:
Formula:
C₉H₁₀N₂O₅
MDL Number:
MFCD00007123
:
226.3 g/mol
EINECS:
210-688-6