4-(2-Chloroethyl)morpholine hydrochloride, Sigma-Aldrich®
Catalog # SIALC42203-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:186.08 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:222-881-2
UN:2923
CAS Number:3647-69-6
Formula:C₆H₁₃Cl₂NO
MDL Number:MFCD00012797