Order Entry
Germany
ContactUsLinkComponent
4-(2-Chloroethyl)morpholine hydrochloride for synthesis, Sigma-Aldrich®
Toxic
Corrosive
4-(2-Chloroethyl)morpholine hydrochloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Corrosive
4-(2-Chloroethyl)morpholine hydrochloride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    182 - 184 °C (decomposition)
  • Molar Mass:
    186.08 g/mol
  • pH:
    4.5 - 5 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    365 °C

Specification Test Results

Related Information