

4-(2-Chloroethyl)morpholine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3647-69-6
:
Formula:C₆H₁₃Cl₂NO
MDL Number:MFCD00012797
:186.08 g/mol
Melting Pt:182…185 °C
Storage Temperature:Ambient
EINECS:222-881-2
UN:2923
ADR:8,III
:GHS06
:GHS05
Specification Test Results
- Appearance:White to beige Hygroscopic crystalline powder
- Infrared spectrum:Conforms
- Melting point:182 to 185 °C (on dry substance)
- Loss on drying:≤1 % (80 °C)
- Titration with HClO4:≥98.5 % (On dry substance)