Tantalum standard solution, 1000 mg/L Ta in 1 M hydrofluoric acid (from TaCl₅) standard for AAS
Catalog # KRAF03520.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1 M HF
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Tantalum
- Symbol:Ta
Specifications
About this item
Certifications: Traceable to NIST