Rhenium standard solution, 1000 mg/L Re in 1 M nitric acid (from HReO₄) standard for AAS
Catalog # KRAF14139.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1 M HNO₃
- Pack type:Plastic bottle
- Pk:0,1 L
- Element:Rhenium
- Symbol:Re
Specifications
About this item
Certifications: Traceable to NIST