Rhenium standard solution, 1000 mg/L Re in 1 M nitric acid (from Re) standard for ICP
Catalog # KRAF03835.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Rhenium
- Symbol:Re
Specifications
About this item
Certifications: Traceable to NIST