Hafnium standard solution, 1000 mg/L Hf in 1 M hydrochloric acid (from HfOCl?) standard for ICP
Catalog # KRAF03724.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:1 M HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Hafnium
- Symbol:Hf
Specifications
About this item
Certifications: Traceable to NIST