Order Entry
Germany
ContactUsLinkComponent
 
Niobium standard solution, 1000 mg/L Nb in 1 M hydrofluoric acid (from NbCl₅) standard for ICP
  KRAF03732.0000
 :  
Niobium standard solution, 1000 mg/L Nb in 1 M hydrofluoric acid (from NbCl₅) standard for ICP
  KRAF03732.0000
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Niobium
  • Symbol:
    Nb

 

 

 : Traceable to NIST