Germanium standard solution, 1000 mg/L Ge in 0,5 M nitric acid with hydrofluoric acid (max. 1%) (de Ge) standard for ICP
Catalog # KRAF03823.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Germanium
- Symbol:Ge
Specifications
About this item
Certifications: Traceable to NIST