Naphthalene-[D8] 2000 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERYA20905100MB
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,1 mL
- CAS No.:1146-65-2
- Description:Naphthalene-[D8] 2,000 µg/ml in tert-butyl methyl ether
- Formula:C₁₀D₈
- Description:Naphthalene-[D8]