Order Entry
Germany
ContactUsLinkComponent
 
Sulphur standard solution, 5000 mg/L S in water (from (NH₄)₂S₄) standard for AAS
  KRAF02571.2000
 :  
Sulphur standard solution, 5000 mg/L S in water (from (NH₄)₂S₄) standard for AAS
  KRAF02571.2000
 :  

 

  • Concentration:
    5000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Sulphur
  • Symbol:
    S

 

 

 : Traceable to NIST