

Bis(2-chloroethyl)ammonium chloride 98%
Catalog # ACRO106321000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:210 to 215 °C
- Titration with HClO4:≥97.5 %
Specification Test Results
About this item
:
:178.49 g/mol
Melting Pt:210…215 °C
Storage Temperature:Ambient
EINECS:212-479-5
UN:3261
ADR:8,II
:GHS08
:GHS05
CAS Number:821-48-7
Formula:C₄H₁₀Cl₃N
MDL Number:MFCD00012515